missing translation for 'onlineSavingsMsg'
Learn More
Learn More
bis(8-Quinolinolato-KappaN1,KappaO8)-Copper (>90%), TRC
Specifications
Specifications
| CAS | 10380-28-6 |
| Molecular Formula | C18 H12 Cu N2 O2 |
| Quantity | 1 g |
| Synonym | Oxine Copper, Copper, bis(8-quinolinolato-κN1,κO8)- (9CI, ACI), Bis(8-quinolinolato-κN1,κO8)copper (ACI), Copper, bis(8-quinolinolato)- (6CI, 8CI), Copper, bis(8-quinolinolato-N1,O8)- (ZCI), 8-Hydroxyquinoline copper salt, 8-Hydroxyquinoline copper(2+) salt, Arbrex 805, Betaxate, Bioquin, Bis(8-hydroxyquinolinato)copper, Bis(8-oxyquinoline)copper, Bis(8-quinolinolato)copper, Bis(8-quinolinolato)copper(II), Copper 8-hydroxyquinolate, Copper 8-hydroxyquinolinate, Copper 8-hydroxyquinoline, Copper 8-quinolate, Copper 8-quinolinol, Copper 8-quinolinolate, Copper 8-quinolinolate (1:2), Copper bis(8-hydroxyquinolinate), Copper Clear, Copper dioxinate, Copper hydroxyquinolate, Copper hydroxyquinolinate, Copper oxinate, Copper oxyquinolate, Copper oxyquinoline, Copper quinolate, Copper quinolinolate, Copper(2+) oxinate, Copper(II) bis(8-quinolinolate), Copper(II) oxinate, Copper-oxine, Cu 8, Cu 8Q, Cunilate 2174, Cunilate 2419, Cunilate 2419-75, Cupric 8-hydroxyquinolate, CUPRIC 8-OXYQUINOLINE, Cupric 8-quinolinolate, Cuprinol Dispersion AQ, Cuproquin, Fennotox 45, Milmer 1, NSC 7720, Nylate 10, Oxidong, Oxine-copper, Oxyquinoline copper, PEI 273, Penetraat, PQ 357, PQ 57, PQ 8, PQ 8 (fungicide), Quin 20, Quinolate, Quinolate 15, Quinolate 20, Timtex, Timtex 17, Timtex AQS, Yukilate CD 50 |
| InChI Formula | InChI=1S/2C9H7NO.Cu/c2*11-8-5-1-3-7-4-2-6-10-9(7)8;/h2*1-6,11H;/q;;+2/p-2 |
| SMILES | O([Cu]Oc1cccc2cccnc12)c3cccc4cccnc34 |
| IUPAC Name | di(quinolin-8-yloxy)copper |
| Molecular Weight (g/mol) | 351.85 |
| Formula Weight | 351.0195 |
| Chemical Name or Material | Oxine-Copper |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?