missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyphenyl) carbonate, 98%, Thermo Scientific™

Catalog Number 11363386
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
553-17-3
C15H14O5
274.272
MFCD00025723
ORUJFMPWKPVXLZ-UHFFFAOYSA-N
guaiacol carbonate, bis 2-methoxyphenyl carbonate, duotal, tussophob, guaiacyl carbonate, di-o-methoxyphenyl carbonate, bis-o-methoxyphenyl carbonate, carbonic acid bis 2-methoxyphenyl ester, guaiacol carbonic acid neutral ester, unii-q71xpq6r29
11104
bis(2-methoxyphenyl) carbonate
COC1=CC=CC=C1OC(=O)OC2=CC=CC=C2OC
This item is not returnable. View return policy
553-17-3
C15H14O5
274.272
MFCD00025723
ORUJFMPWKPVXLZ-UHFFFAOYSA-N
guaiacol carbonate, bis 2-methoxyphenyl carbonate, duotal, tussophob, guaiacyl carbonate, di-o-methoxyphenyl carbonate, bis-o-methoxyphenyl carbonate, carbonic acid bis 2-methoxyphenyl ester, guaiacol carbonic acid neutral ester, unii-q71xpq6r29
11104
bis(2-methoxyphenyl) carbonate
COC1=CC=CC=C1OC(=O)OC2=CC=CC=C2OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 86°C to 88°C
Quantity 5 g
Beilstein 2472687
Formula Weight 274.28
Percent Purity 98%
Chemical Name or Material Bis(2-methoxyphenyl) carbonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'