Learn More
Bis(2-methoxyethyl) Phthalate, TRC
Product Code. 30179078
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179078 10 g 10g
30179456 1 g 1g
30179675 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179078 Supplier Toronto Research Chemicals Supplier No. TRCB48751010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 117-82-8
Molecular Formula C14 H18 O6
Molecular Weight (g/mol) 282.29
Synonym 1,2-Benzenedicarboxylic acid, 1,2-bis(2-methoxyethyl) ester, 1,2-Benzenedicarboxylic acid, bis(2-methoxyethyl) ester (9CI), Phthalic acid, bis(2-methoxyethyl) ester (6CI,7CI,8CI), Ethanol, 2-methoxy-, phthalate (2:1) (8CI), 2-Methoxyethyl phthalate, Bis(2-methoxyethyl) phthalate, Bis(2-methoxyethyl)ester 1,2-benzenedicarboxylic acid, Bis(methoxyethyl) phthalate, Di(2-methoxyethyl) phthalate, Dimethyl glycol phthalate, KC 988, Kesscoflex MCP, Methyl glycol phthalate, NSC 2147
IUPAC Name bis(2-methoxyethyl) benzene-1,2-dicarboxylate
SMILES COCCOC(=O)c1ccccc1C(=O)OCCOC
Specifica
Quantity 10 g
InChI Formula InChI=1S/C14H18O6/c1-17-7-9-19-13(15)11-5-3-4-6-12(11)14(16)20-10-8-18-2/h3-6H,7-10H2,1-2H3
Formula Weight 282.1103
Chemical Name or Material Phthalic acid, bis-methylglycol ester
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.