Learn More
Bis(2-ethylhexyl) Azelate, TRC
Product Code. 30132977
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132977 10 g 10g
30003232 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30132977 Supplier Toronto Research Chemicals Supplier No. TRCB43382010G
 Restricted Item
Estimated Shipment: 12-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103-24-2
Molecular Formula C25 H48 O4
Molecular Weight (g/mol) 412.65
Synonym Azelaic acid, bis(2-ethylhexyl) ester (6CI,7CI,8CI), Nonanedioic acid, bis(2-ethylhexyl) ester (9CI), Azelaic acid di(2-ethylhexyl) ester, Bis(2-ethylhexyl) azelate, Bis(2-ethylhexyl) nonanedioate, DOZ, Di-2-ethylhexyl azelate, Dioctyl azelate, Dioctyl nonanedioate, Edenol 9058, Emery 2958, Emolien 2986, Nonanedioic acid di(2-ethylhexyl) ester, Octyl azelate, Plastolein 9058, Plastolein 9058DOZ, Sansocizer DOZ, Staflex DOX, Truflex DOX, Nonanedioic acid, 1,9-bis(2-ethylhexyl) ester
IUPAC Name bis(2-ethylhexyl) nonanedioate
SMILES CCCCC(CC)COC(=O)CCCCCCCC(=O)OCC(CC)CCCC
Specifications
Quantity 10 g
InChI Formula InChI=1S/C25H48O4/c1-5-9-16-22(7-3)20-28-24(26)18-14-12-11-13-15-19-25(27)29-21-23(8-4)17-10-6-2/h22-23H,5-21H2,1-4H3
Formula Weight 412.3553
Chemical Name or Material Bis(2-ethylhexyl) Azelate