missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,6-hydroxymethyl)dioxane (Mixture of Diastereomers), TRC

Product Code. 30010457
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010457 25 mg 25mg
30005817 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010457 Supplier Toronto Research Chemicals Supplier No. TRCB44530525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54120-69-3
Molecular Formula C6 H12 O4
Molecular Weight (g/mol) 148.16
Synonym 1,4-Dioxane-2,6-dimethanol, p-Dioxane-2,6-dimethanol (6CI), 1,4-Dioxan-2,6-dimethanol, Bis(2,6-hydroxymethyl)dioxane
IUPAC Name [6-(hydroxymethyl)-1,4-dioxan-2-yl]methanol
SMILES OCC1COCC(CO)O1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H12O4/c7-1-5-3-9-4-6(2-8)10-5/h5-8H,1-4H2
Formula Weight 148.0736
Percent Purity >95%
Chemical Name or Material Bis(2,6-hydroxymethyl)dioxane(Mixture of Diastereomers)
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.