Learn More
(+)-Biotin 4-Amidobenzoic Acid, Sodium Salt, TRC
Spezifikation
Spezifikation
| Chemical Name or Material | (+)-Biotin 4-Amidobenzoic Acid, Sodium Salt |
| CAS | 102418-74-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 H20 N3 O4 S . Na |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C17H21N3O4S.Na/c21-14(18-11-7-5-10(6-8-11)16(22)23)4-2-1-3-13-15-12(9-25-13)19-17(24)20-15;/h5-8,12-13,15H,1-4,9H2,(H,18,21)(H,22,23)(H2,19,20,24);/q;+1/p-1/t12?,13-,15?;/m0./s1 |
| SMILES | [Na+].[O-]C(=O)c1ccc(NC(=O)CCCC[C@@H]2SCC3NC(=O)NC23)cc1 |
| IUPAC Name | sodium;4-[5-[(4S)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]benzoate |
| Molecular Weight (g/mol) | 385.41 |
| Formula Weight | 385.1072 |
| Mehr anzeigen |