Learn More
Intelsius™ BioTherm™ Dry Ice Shipper
Product Code. 17484602
Encompass_Preferred
Change view
Click to view available options
Capacity (Metric):
6.6 L
32.7 L
Weight (Metric):
Dry Ice: 13.6 kg
Dry Ice: 3.9 kg
Holds Temp., Hours:
72 hr.
96+ hr.
Unit Size:
Pack of 24
Pack of 60
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Weight (Metric) Holds Temp., Hours unitSize
17484602 6.6 L Dry Ice: 3.9 kg 72 hr. Pack of 60
17454602 32.7 L Dry Ice: 13.6 kg 96+ hr. Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17484602 Supplier Intelsius™ Supplier No. BT074
Add to basket
Add to basket
The BioTherm™ Dry Ice Shipper is a UN3373 complaint dry ice shipper designed to offer protection when transporting deep frozen samples.
  • The BioTherm Dry ice range features high performance EPS insulation which maximises the available payload space and temperature stability when used with dry ice as a coolant.
  • All the necessary shipping labelling to achieve compliance during transportation is provided.
Specifications
Dimensions (L x W x H) 190 x 190 x 185 mm (7.4 x 7.4 x 7.2 in.)
For Use With (Equipment) EPS 7
Weight (Metric) Dry Ice: 3.9 kg
Holds Temp., Hours 72 hr.
Type Insulated Dry Ice Carrier
Capacity (Metric) 6.6 L
Holds 600 mL