Learn More
Gilac™ Biobased 22 Cycle Tumblers
Product Code. 17918671
Change view
Click to view available options
Capacity (Metric):
0.22 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17918671 0.22 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17918671 Supplier Gilac™ Supplier No. G627225
  • ONLINE EXCLUSIVE PRICE
    11.75 EUR
    7.94 EUR / Pack of 1
    Save 3.81 EUR
    32% Off
Estimated Shipment: 29-12-2026
Biobased 22 Cycle Tumblers are reusable biobased plastic tumblers with outstanding durability even in intensive cycle use.
  • Reusable biobased plastic tumblers.
  • Outstanding durability even in intensive cycle use.
  • Ideal for institutions.
  • Pack of 10.
  • Biobased tumblers are white and dye-free.
  • Biobased material is made from natural renewable resources instead of fossil resources.
  • As a responsible French manufacturer, GILAC is now offering alternatives to standard materials, whilst maintaining an optimal level of quality and food safety.
Specifications
Material HDPE
Capacity (Metric) 0.22 L
Capacity (English) 7.4 oz.
Quantity 10 Pk.
Product Type Tumbler Cup