Learn More
Hirschmann™ BIO-SILICO™ Silicone Stoppers
Product Code. 10007792
Change view
Click to view available options
Diameter (Metric) Inner:
11-14 mm
15-18 mm
18-21 mm
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10007792 18-21 mm Pack of 10
10057512 11-14 mm Pack of 10
10186780 15-18 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10007792 Supplier Hirschmann™ Supplier No. 8905922
  • ONLINE EXCLUSIVE PRICE
    86.21 EUR
    80.10 EUR / Pack of 10
    Save 6.11 EUR
    7% Off
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Excellent for the cultivation of aerobic microorganisms. Specially processed culture plugs with coarse bubbles for preparation, filling and sterilization of culture media.
  • Coarse pore structure
  • Good air permeability
  • Chemically and temperature resistant
  • Liquid repellent and reusable
Specifications
Description BIO-SILICO, N22
Diameter (Metric) Inner 18-21 mm
For Use With (Application) For The Preparation And Sterilization Of Culture Media
Length (Metric) Ground Zone 30 mm
Length (Metric) Sleeve 18 mm
Quantity 10/Pack
Color Orange
Diameter (Metric) Bottom 18 mm
Diameter (Metric) Top 24 mm
Material Silicone
Product Type Silicone Stopper