Learn More
Bilirubin (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | Bilirubin |
| CAS | 635-65-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C33 H36 N4 O6 |
| Quantity | 25 g |
| Synonym | 21H-Biline-8,12-dipropanoic acid, 2,17-diethenyl-1,10,19,22,23,24-hexahydro-3,7,13,18-tetramethyl-1,19-dioxo-, Biline-8,12-dipropionic acid, 1,10,19,22,23,24-hexahydro-2,7,13,17-tetramethyl-1,19-dioxo-3,18-divinyl- (8CI), Hematoidin (6CI), 2,17-Diethenyl-1,10,19,22,23,24-hexahydro-3,7,13,18-tetramethyl-1,19-dioxo-21H-biline-8,12-dipropanoic acid, (4Z,15Z)-Bilirubin IXα, (Z,Z)-Bilirubin, (Z,Z)-Bilirubin IXα, Bilirubin, Bilirubin IXα, Cholerythrin, NSC 26685 |
| InChI Formula | InChI=1S/C33H36N4O6/c1-7-20-19(6)32(42)37-27(20)14-25-18(5)23(10-12-31(40)41)29(35-25)15-28-22(9-11-30(38)39)17(4)24(34-28)13-26-16(3)21(8-2)33(43)36-26/h7-8,13-14,34-35H,1-2,9-12,15H2,3-6H3,(H,36,43)(H,37,42)(H,38,39)(H,40,41)/b26-13-,27-14- |
| SMILES | CC1=C(C=C)\C(=C\c2[nH]c(Cc3[nH]c(\C=C\4/NC(=O)C(=C4C)C=C)c(C)c3CCC(=O)O)c(CCC(=O)O)c2C)\NC1=O |
| IUPAC Name | 3-[2-[[3-(2-carboxyethyl)-5-[(Z)-(3-ethenyl-4-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-2-yl]methyl]-5-[(Z)-(4-ethenyl-3-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-3-yl]propanoic acid |
| Molecular Weight (g/mol) | 584.662 |
| Show More |