missing translation for 'onlineSavingsMsg'
Learn More

Bicyclopyrone, TRC

Product Code. 30138696
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138696 5 mg 5mg
18012740 2.5 mg 2.5mg
18041500 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138696 Supplier Toronto Research Chemicals Supplier No. TRCB3821005MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352010-68-5
Molecular Formula C19 H20 F3 N O5
Molecular Weight (g/mol) 399.36
Synonym 4-Hydroxy-3-[[2-[(2-methoxyethoxy)methyl]-6-(trifluoromethyl)-3-pyridinyl]carbonyl]bicyclo[3.2.1]oct-3-en-2-one, Bicyclopyrone
IUPAC Name 2-hydroxy-3-[2-(2-methoxyethoxymethyl)-6-(trifluoromethyl)pyridine-3-carbonyl]bicyclo[3.2.1]oct-2-en-4-one
SMILES COCCOCc1nc(ccc1C(=O)C2=C(O)C3CCC(C3)C2=O)C(F)(F)F

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H20F3NO5/c1-27-6-7-28-9-13-12(4-5-14(23-13)19(20,21)22)18(26)15-16(24)10-2-3-11(8-10)17(15)25/h4-5,10-11,24H,2-3,6-9H2,1H3
Formula Weight 399.1294
Percent Purity >95%
Chemical Name or Material Bicyclopyrone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.