missing translation for 'onlineSavingsMsg'
Learn More

Bictegravir, MedChemExpress

Product Code. 16451709
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451709 1 mg 1mg
16447418 10 mg 10mg
16439897 5 mg 5mg
16437396 100 mg 100mg
16413447 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16451709 Supplier Medchem Express Supplier No. HY17605

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Bictegravir (GS-9883) is a potent inhibitor of HIV-1 integrase with an IC50 of 7.5 nM.

Bictegravir (GS-9883) is a potent inhibitor of HIV-1 integrase with an IC50 of 7.5 nM.

Specifications

Chemical Name or Material Bictegravir
CAS 1611493-60-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H18F3N3O5
Quantity 1 mg
Synonym GS-9883
Solubility Information DMSO : 83.3 mg/mL (185.37 mM; Need ultrasonic and warming)
SMILES O=C1N([C@](CC2)([H])C[C@]2([H])O3)[C@@]3([H])CN(C=C(C(NCC(C(F)=CC(F)=C4)=C4F)=O)C5=O)C1=C5O
Molecular Weight (g/mol) 449.38
Formula Weight 449.38
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.