missing translation for 'onlineSavingsMsg'
Learn More

Bicalutamide, TRC

Product Code. 30133724
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Unit Size:
100mg
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133724 2.5 g 2.5g
30137740 500 mg 500mg
30041291 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133724 Supplier Toronto Research Chemicals Supplier No. TRCB3820002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90357-06-5
Molecular Formula C18 H14 F4 N2 O4 S
Molecular Weight (g/mol) 430.37
Synonym N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide, (±)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide, (±)-4'-Cyano-α,α,α-trifluoro-3-[(p-fluorophenyl)sulfonyl]-2-methyl-m-lactotoluidide, Bicalutamide, Casodex, Cosudex, ICI 176334, ZD 176334
IUPAC Name N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanamide
SMILES CC(O)(CS(=O)(=O)c1ccc(F)cc1)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C18H14F4N2O4S/c1-17(26,10-29(27,28)14-6-3-12(19)4-7-14)16(25)24-13-5-2-11(9-23)15(8-13)18(20,21)22/h2-8,26H,10H2,1H3,(H,24,25)
Formula Weight 430.061
Chemical Name or Material Bicalutamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.