missing translation for 'onlineSavingsMsg'
Learn More

BIBR 1087 SE, MedChemExpress

Product Code. 16440469
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16440469 10 mg 10mg
16472668 5 mg 5mg
16464126 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16440469 Supplier Medchem Express Supplier No. HYW004360
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BIBR 1087 SE is an intermediate metabolite of dabigatran etexilate.

BIBR 1087 SE is an intermediate metabolite of dabigatran etexilate.

Specifications

Chemical Name or Material BIBR 1087 SE
CAS 212321-78-3
Color White
Purity Grade Notes Research
Molecular Formula C32H37N7O5
Quantity 10 mg
Synonym Desethyl Dabigatran Etexilate
Solubility Information DMSO : ≥ 100 mg/mL (166.76 mM)
SMILES N=C(C1=CC=C(C=C1)NCC2=NC3=CC(C(N(CCC(O)=O)C4=NC=CC=C4)=O)=CC=C3N2C)NC(OCCCCCC)=O
Molecular Weight (g/mol) 599.68
Formula Weight 599.68
Percent Purity 96.57%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.