Learn More
Keystone Cleanroom Products BHD Autoclavable Wrapper
BIOPHARMAGREENER_CHOICE
Product Code. 17798177 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Length (English):
1.5 in.
2 in.
3 in.
4 in.
6 in.
Width (English):
1.5 in.
2 in.
3 in.
4 in.
6 in.
Unit Size:
Pack of 3000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Width (English) unitSize
17798177 1.5 in. 1.5 in. Pack of 3000
17708187 2 in. 2 in. Pack of 3000
17718187 3 in. 3 in. Pack of 3000
17728187 4 in. 4 in. Pack of 3000
17738187 6 in. 6 in. Pack of 3000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17798177 Supplier Keystone Cleanroom Products Supplier No. BHDSHEET01.5X01.5
  • ONLINE EXCLUSIVE PRICE
    812.00 EUR
    687.07 EUR / Pack of 3000
    Save 124.93 EUR
    15% Off
View all promotions
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
When secured with autoclave tape or elasticized bands, these BHD Autoclavable Wrappers provide an adequate microbial barrier from external sources of contamination.
  • Designed for steam sterilization
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
  • Sufficient microbial barrier
  • Resists tearing and punctures
  • Does not contain any natural latex rubber
  • Processed and packaged in ISO Certified Cleanrooms
  • Certificate of Conformance included with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance
Closure Type None
Color Blue
Disposable Single-use
For Use With (Application) Gamma, EtO, or Steam Sterilization
Material Cellulose
Sterility Non-sterile
Latex Free Yes
Temperature Range (Metric) Up to 135°C
Length (English) 1.5 in.
Width (English) 1.5 in.
Product Type Sterilization Wrap (Heavy-duty)
Length (Metric) 3.8 cm
Width (Metric) 3.8 cm