Learn More
Scientific Industries BG Replacement Top with Screw
Product Code. 17281752 Shop All Ohaus Products
Change view
Click to view available options
For Use With (Equipment):
Bead Genie
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17281752 Bead Genie Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17281752 Supplier Scientific Industries Supplier No. 83041453
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
BG Replacement Top with Screw is a replacement clamping disk with screw that is required to clamp all holders to the Bead Genie.
  • Replacement clamping disk with screw required to clamp holders to Bead Genie
TRUSTED_SUSTAINABILITY
Especificaciones
Description BG Replacement Top with Screw
For Use With (Equipment) Bead Genie
Model SI-B150
Packaging Quantity 1 Ea.
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.