missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Bexarotene
GREENER_CHOICE

Product Code. 18731348
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18731348 10 mg 10mg
18768788 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18731348 Supplier Tocris Bioscience™ Supplier No. 5819/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective RXR agonist

Bexarotene is a potent and selective RXR agonist (EC50 values are 24, 25 and 33 nM for RXRβ, RXRγ and RXRα, respectively). Exhibits >300-fold selectivity for RXR over RAR receptors. Inhibits tumor cell invasion and migration in vitro, and inhibits metastasis and angiogenesis in mouse models. Also promotes brown adipogenic differentiation and induces brown adipogenic reprogramming from myoblasts.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[1-(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)ethenyl]benzoic acid
CAS 153559-49-0
Synonym LGD 1069
Quantity 10 mg
Target Retinoid X Receptor Agonists
Molecular Formula C24H28O2
Formula Weight 348.48
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.