missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BETP, MedChemExpress
Description
BETP is an agonist of glucagon-like peptide-1 (GLP-1) receptor, with EC50s of 0.66 and 0.755 μM for human and rat GLP-1 receptor, respectively.
Specifications
Specifications
| Chemical Name or Material | BETP |
| CAS | 1371569-69-5 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C20H17F3N2O2S |
| Quantity | 5 mg |
| Solubility Information | DMSO : 25 mg/mL (61.51 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | FC(C1=CC(C2=CC=CC(OCC3=CC=CC=C3)=C2)=NC(S(CC)=O)=N1)(F)F |
| Molecular Weight (g/mol) | 406.42 |
| Formula Weight | 406.42 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?