missing translation for 'onlineSavingsMsg'
Learn More

BETP, MedChemExpress

Product Code. 16493858
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493858 10 mg 10mg
16472876 25 mg 25mg
16466587 5 mg 5mg
16424896 100 mg 100mg
16420237 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493858 Supplier Medchem Express Supplier No. HY103546

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BETP is an agonist of glucagon-like peptide-1 (GLP-1) receptor, with EC50s of 0.66 and 0.755 μM for human and rat GLP-1 receptor, respectively.

BETP is an agonist of glucagon-like peptide-1 (GLP-1) receptor, with EC50s of 0.66 and 0.755 μM for human and rat GLP-1 receptor, respectively.

Specifications

Chemical Name or Material BETP
CAS 1371569-69-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H17F3N2O2S
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (61.51 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES FC(C1=CC(C2=CC=CC(OCC3=CC=CC=C3)=C2)=NC(S(CC)=O)=N1)(F)F
Molecular Weight (g/mol) 406.42
Formula Weight 406.42
Percent Purity 98.3%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.