Learn More
Beta-Naphthylcarbinol, TRC
Product Code. 30480384
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30480384 5 g
30474624 10 g
30472676 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30480384 Supplier Toronto Research Chemicals Supplier No. TRCN3456455G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1592-38-7
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.2
Synonym 2-(Hydroxymethyl)naphthalene, 2-Naphthylcarbinol, 2-Naphthylmethanol, 2-Naphthylmethyl Alcohol, NSC 408615, Naphthalen-2-ylmethanol, beta-(Hydroxymethyl)naphthalene, 2-Naphthalenemethanol
IUPAC Name naphthalen-2-ylmethanol
SMILES OCc1ccc2ccccc2c1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C11H10O/c12-8-9-5-6-10-3-1-2-4-11(10)7-9/h1-7,12H,8H2
Formula Weight 158.0732
Chemical Name or Material beta-Naphthylcarbinol