Learn More
Fisherbrand™ Beta Mini Radiation Storage Containers
Product Code. 12397097 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Height (Metric):
55mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12397097 55mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12397097
Estimated Shipment: 05-04-2027
Add to basket
Add to basket
Ideal solution for storing radioactive materials, to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes.
  • Beta – Mini (5.5 x 8.5 x 8.5cm)
  • For storing radioactive materials, to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, and the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Specifications
For Use With (Application) To hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
Height (Metric) 55mm
Holds Microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
Product Type Radiation Storage Box