Learn More
Fisherbrand™ Radiation Safety Storage Box Inserts
Product Code. 12337167 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Tube Size:
8 x 1.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tube Size Unit Size
12337167 8 x 1.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12337167
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
Manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes; these serve as an ideal solution for short-term storage of radioactive waste or radioisotopes.
  • Insert, 2 x Falcon™ tubes, 8 x 1.5mL tubes, 160 x 80 x 40mm
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, and the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Specifications
For Use With (Equipment) For Maxi Boxes
Height (Metric) 40mm
Product Type Insert
Tube Size 8 x 1.5 mL
Width (Metric) 160mm