Learn More
Fisherbrand™ Radiation Safety Storage Bins
Product Code. 12337107 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Height (Metric):
130mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12337107 130mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12337107
Estimated Shipment: 09-04-2027
Add to basket
Add to basket
Radiation boxes, with hinged lid and central swivel covers, that are ideal for protection against Beta or Gamma radiation.
  • Beta – 1L – (13 x 10 x 8cm), use with bag CSR-BAG1
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, and the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Specifications
For Use With (Application) For storing radioactive materials, To hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
Height (Metric) 130mm
Holds Microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
Product Type Radiation Storage Box
Capacity 1 L