Learn More
Beryllium Standard: Be @ 10000 μg/mL in 5% HNO3, VHG™
Product Code. 17957499
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17957499 100 mL 100mL
17948249 250 mL 250mL
17938979 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17957499 Supplier VHG Labs Supplier No. VHGTBEN100
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Beryllium Standard: Be @ 10000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Beryllium
CAS 7440-41-7
Methods ICP/ICP-MS
Analyte or Component Names Beryllium
Shipping Condition Room Temperature
Molecular Formula Be
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
Synonym Glucinium, Beryllium dust, Beryllium metal, Beryllium powder
InChI Formula InChI=1S/Be
SMILES [Be]
IUPAC Name beryllium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 9.012
Quantity 100 mL
Formula Weight 9.012 g/mol