Learn More
Bergaptol, TRC
Product Code. 30134883
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134883 250 mg 250mg
30011136 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30134883 Supplier Toronto Research Chemicals Supplier No. TRCB318510250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 486-60-2
Molecular Formula C11 H6 O4
Molecular Weight (g/mol) 202.16
Synonym 4-Hydroxy-7H-furo[3,2-g][1]benzopyran-7-one, 4-Hydroxybergapten, 5-Hydroxy-6,7-furanocoumarin, 5-Hydroxypsoralen, NSC 341958;
IUPAC Name 4-hydroxyfuro[3,2-g]chromen-7-one
SMILES Oc1c2C=CC(=O)Oc2cc3occc13
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C11H6O4/c12-10-2-1-6-9(15-10)5-8-7(11(6)13)3-4-14-8/h1-5,13H
Formula Weight 202.0266
Chemical Name or Material Bergaptol