Learn More
Bergapten, TRC
Product Code. 30009307
Change view
Click to view available options
Quantity:
50 mg
250 mg
10 g
Unit Size:
10g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009307 10 g 10g
30003973 250 mg 250mg
17954965 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009307 Supplier Toronto Research Chemicals Supplier No. TRCB31850010G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 484-20-8
Molecular Formula C12 H8 O4
Molecular Weight (g/mol) 216.19
Synonym 7H-Furo[3,2-g][1]benzopyran-7-one, 4-methoxy-, 4-Methoxy-7H-furo[3,2-g][1]benzopyran-7-one, 5-MOP, 5-Methoxy-6,7-furanocoumarin, 5-Methoxypsoralen, 8-Demethoxyisoimpinellin, Bergaptan, Bergapten, Bergaptene, Geralen, Heraclin, Majudin, NSC 95437, Psoraderm
IUPAC Name 4-methoxyfuro[3,2-g]chromen-7-one
SMILES COc1c2C=CC(=O)Oc2cc3occc13
Specifications
Quantity 10 g
InChI Formula InChI=1S/C12H8O4/c1-14-12-7-2-3-11(13)16-10(7)6-9-8(12)4-5-15-9/h2-6H,1H3
Formula Weight 216.0423
Chemical Name or Material Bergapten