Learn More
Beraprost Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Beraprost sodium salt |
| CAS | 88475-69-8 |
| Molecular Formula | C24 H29 O5 . Na |
| Quantity | 10 mg |
| Synonym | 1H-Cyclopenta[b]benzofuran-5-butanoic acid, 2,3,3a,8b-tetrahydro-2-hydroxy-1-(3-hydroxy-4-methyl-1-octen-6-yn-1-yl)-, sodium salt (1:1), 1H-Cyclopenta[b]benzofuran-5-butanoic acid, 2,3,3a,8b-tetrahydro-2-hydroxy-1-(3-hydroxy-4-methyl-1-octen-6-ynyl)-, monosodium salt (9CI), Beraprost sodium, Beraprost sodium salt, Berasus, Careload, Dorner, MDL 201129, ML 1129, Procylin, TRK 100, dl-4-[(1R,2R,3aS,8bS)-2,3,3a,8b-Tetrahydro-2-hydroxy-[(3S,4RS)-3-hydroxy-4-methyl-oct-6-yne-(E)-1-enyl]-5-cyclopenta[b]benzofuranyl]butanoic Acid Sodium |
| InChI Formula | InChI=1S/C24H30O5.Na/c1-3-4-7-15(2)19(25)13-12-17-20(26)14-21-23(17)18-10-5-8-16(24(18)29-21)9-6-11-22(27)28;/h5,8,10,12-13,15,17,19-21,23,25-26H,6-7,9,11,14H2,1-2H3,(H,27,28);/q;+1/p-1/b13-12+;/t15?,17-,19+,20+,21-,23-;/m0./s1 |
| SMILES | [Na+].CC#CCC(C)[C@H](O)\C=C\[C@H]1[C@H](O)C[C@@H]2Oc3c(CCCC(=O)[O-])cccc3[C@H]12 |
| IUPAC Name | sodium;4-[(1R,2R,3aS,8bS)-2-hydroxy-1-[(E,3S)-3-hydroxy-4-methyloct-1-en-6-ynyl]-2,3,3a,8b-tetrahydro-1H-cyclopenta[b][1]benzofuran-5-yl]butanoate |
| Molecular Weight (g/mol) | 420.47 |
| Formula Weight | 420.1913 |