Learn More
Beperidium iodide, MedChemExpress
Product Code. 16465547
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465547 5 mg 5mg
16422808 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16465547 Supplier Medchem Express Supplier No. HY100152
 Restricted Item
Estimated Shipment: 30-11-2026
MedChemExpress Beperidium iodide shows a competitive antagonistic effect against acetylcholine receptor with a pA2 of 7.93.
Beperidium iodide shows a competitive antagonistic effect against acetylcholine receptor with a pA2 of 7.93.
Specifications
Chemical Name or Material Beperidium iodide
CAS 86434-57-3
Color White
Purity Grade Notes Research
Molecular Formula C23H34IN3O3
Quantity 5 mg
Synonym SX 810
Solubility Information DMSO : ≥ 100 mg/mL (189.60 mM)
SMILES C[N+]1(CC)CCC(OC(C(C2=NOC3=C2C=CC=C3)N4CCCCCC4)=O)CC1.[I-]
Molecular Weight (g/mol) 527.44
Formula Weight 527.44
Percent Purity 99.79%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Vis mere Vis mindre
Research purposes only
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.