missing translation for 'onlineSavingsMsg'
Learn More

Benzylacetophenone, MP Biomedicals

Product Code. 11468605 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468605 10 g 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11468605 Supplier MP Biomedicals Supplier No. 0522431450

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1083-30-3
Molecular Formula C15H14O
Quantity 10 g
Synonym dihydrochalcone, 3-phenylpropiophenone, benzylacetophenone, hydrochalcone, 1,3-diphenyl-1-propanone, beta-phenylpropiophenone, hydrocinnamophenone, 1,3-diphenyl-1-oxopropane, phenethyl phenyl ketone, phenyl phenethyl ketone
InChI Key QGGZBXOADPVUPN-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)CCC(=O)C2=CC=CC=C2
IUPAC Name 1,3-diphenylpropan-1-one
Molecular Weight (g/mol) 210.276
PubChem CID 64802
ChEBI CHEBI:71231
Formula Weight 210.29
Chemical Name or Material Benzylacetophenone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.