Learn More
Benzyl (2S)-3-Methyl-2-[pentanoyl[[2'-[2-(triphenylmethyl)tetrazol-5-yl]biphenyl-4-yl]methyl]amino]butanoate, Mikromol™
Description
Benzyl (2S)-3-Methyl-2-[pentanoyl[[2'-[2-(triphenylmethyl)tetrazol-5-yl]biphenyl-4-yl]methyl]amino]butanoate
Specifications
Specifications
| Chemical Name or Material | Benzyl (2S)-3-Methyl-2-[pentanoyl[(2'-[2-(triphenylmethyl)tetrazol-5-yl]biphenyl-4-yl)methyl]amino]butanoate |
| CAS | 2459446-33-2 |
| Analyte or Component Names | Benzyl (2S)-3-Methyl-2-[pentanoyl[(2'-[2-(triphenylmethyl)tetrazol-5-yl]biphenyl-4-yl)methyl]amino]butanoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C50 H49 N5 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzyl (2S)-3-Methyl-2-[pentanoyl[[2'-[2-(triphenylmethyl)tetrazol-5-yl]biphenyl-4-yl]methyl]amino]butanoate, N2-Tritylvalsartan Benzyl Ester |
| InChI Formula | InChI=1S/C50H49N5O3/c1-4-5-30-46(56)54(47(37(2)3)49(57)58-36-39-20-10-6-11-21-39)35-38-31-33-40(34-32-38)44-28-18-19-29-45(44)48-51-53-55(52-48)50(41-22-12-7-13-23-41,42-24-14-8-15-25-42)43-26-16-9-17-27-43/h6-29,31-34,37,47H,4-5,30,35-36H2,1-3H3/t47-/m0/s1 |
| SMILES | CCCCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnn(n3)C(c4ccccc4)(c5ccccc5)c6ccccc6)[C@@H](C(C)C)C(=O)OCc7ccccc7 |
| IUPAC Name | benzyl (2S)-3-methyl-2-[pentanoyl-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]amino]butanoate |
| Show More |