Learn More
Benzyl 2,3-O-Isopropylidene-Alpha-D-mannopentenofuranoside-6-aldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | Benzyl 2,3-O-Isopropylidene-a-D-mannopentenofuranoside-6-ald |
| CAS | 102854-75-1 |
| Molecular Formula | C15H18O5 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C15H18O5/c1-15(2)19-12-11(8-16)18-14(13(12)20-15)17-9-10-6-4-3-5-7-10/h3-8,11-14H,9H2,1-2H3/t11-,12+,13+,14+/m1/s1 |
| SMILES | CC1(C)O[C@@H]2[C@@H](OCc3ccccc3)O[C@H](C=O)[C@@H]2O1 |
| IUPAC Name | (3aS,4S,6S,6aS)-2,2-dimethyl-4-phenylmethoxy-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxole-6-carbaldehyde |
| Molecular Weight (g/mol) | 278.3 |
| Formula Weight | 278.1154 |