Learn More
Benzyl (1,3-Dihydroxy-2-(hydroxymethyl)propan-2-yl)carbamate, TRC
Specifications
Specifications
| Chemical Name or Material | N-[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]carbamic Acid Phenylmethyl Ester |
| CAS | 102522-48-5 |
| Molecular Formula | C12H17NO5 |
| Quantity | 2.5 g |
| Synonym | N-Carboxybenzyl Tromethamine |
| InChI Formula | InChI=1S/C12H17NO5/c14-7-12(8-15,9-16)13-11(17)18-6-10-4-2-1-3-5-10/h1-5,14-16H,6-9H2,(H,13,17) |
| SMILES | O=C(OCC=1C=CC=CC1)NC(CO)(CO)CO |
| IUPAC Name | benzyl (1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)carbamate |
| Molecular Weight (g/mol) | 255.27 |
| Formula Weight | 255.11 |