missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid Standard, For Elementary Analysis, MilliporeSigma™

Product Code. 15853830
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15853830 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15853830 Supplier Supelco Supplier No. 1001340005
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 65-85-0
Appearance White solid
Quantity 5 g
Ignition Point 570°C
Odor Characteristic odor
Packaging Glass bottle
Physical Form Solid
Molecular Formula C7H6O2
Vapor Pressure 0.001 hPa at 20°C
Synonym benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv
InChI Key WPYMKLBDIGXBTP-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)O
IUPAC Name benzoic acid
Molecular Weight (g/mol) 122.123
PubChem CID 243
ChEBI CHEBI:30746
Formula Weight 122.12
Chemical Name or Material Benzoic Acid Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.