Learn More
Benzo[h]quinoline, 98%
Product Code. 11473388
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473388 5 g 5g
11463388 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11473388 Supplier Thermo Scientific Alfa Aesar Supplier No. A17794.06
 Restricted Item
In Stock
Add to basket
CAS: 230-27-3 | C13H9N | 179.222 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 230-27-3
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.222
MDL Number MFCD00004984
InChI Key WZJYKHNJTSNBHV-UHFFFAOYSA-N
Synonym benzo h quinoline, 7,8-benzoquinoline, 4-azaphenanthrene, 1-naphthoquinoline, benzoquinoline, alpha-naphthoquinoline, alpha-benzoquinoline, 7,8-bnzoquinoline, .alpha.-benzoquinoline
PubChem CID 9191
IUPAC Name benzo[h]quinoline
SMILES C1=CC=C2C(=C1)C=CC3=C2N=CC=C3
Specifications
Melting Point 48°C to 52°C
Boiling Point 233°C (47 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 120249
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material Benzo[h]quinoline