Learn More
Benzil, Sigma-Aldrich™
Product Code. 15868610
Change view
Click to view available options
Quantity:
5 g
250 g
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15868610 250 g 250g
15858610 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15868610 Supplier Sigma Aldrich Supplier No. 8016320250
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 134-81-6
Grade Research
Molecular Weight (g/mol) 210.23
Specifications
Melting Point 95°C
Boiling Point 346°C to 348°C/1013 hPa (Decomposition)
MDL Number MFCD00003080
Quantity 250 g
Synonym Benzil, Dibenzoyl, Benzil