missing translation for 'onlineSavingsMsg'
Learn More

Benzidine, TRC

Product Code. 30040819
Change view
Click to view available options
Quantity:
100 mg
5 g
Unit Size:
100mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040819 5 g 5g
30040459 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30040819 Supplier Toronto Research Chemicals Supplier No. TRCB1210005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-87-5
Molecular Formula C12 H12 N2
Molecular Weight (g/mol) 184.24
Synonym Benzidine (8CI), 4,4'-Bianiline, 4,4'-Biphenyldiamine, 4,4'-Diamino-1,1'-biphenyl, 4,4'-Diaminobenzidine, 4,4'-Diaminobiphenyl, 4,4'-Diaminodiphenyl, 4,4'-Diphenylenediamine, 4-(4-Aminophenyl)aniline, 4'-Amino-[1,1'-biphenyl]-4-ylamine, C.I. Azoic Diazo Component 112, Fast Corinth Base B, NSC 146476, p,p'-Bianiline, p,p'-Diaminobiphenyl, p-Diaminodiphenyl
IUPAC Name 4-(4-aminophenyl)aniline
SMILES Nc1ccc(cc1)c2ccc(N)cc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H12N2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H,13-14H2
Formula Weight 184.1
Percent Purity >95%
Chemical Name or Material 4,4'-Benzidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.