missing translation for 'onlineSavingsMsg'
Learn More

Benzamidine Hydrochloride Hydrate, Sigma-Aldrich™

Product Code. 15860761
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15860761 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15860761 Supplier Sigma-Aldrich Supplier No. 8201220025
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 206752-36-5
Melting Point 80°C to 84°C
MDL Number MFCD00066285
Quantity 25 g
Synonym Benzamidine hydrochloride hydrate, Benzamidine hydrochloride hydrate
Molecular Weight (g/mol) 120.15 (Anhydrous Free Base Basis)
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.