missing translation for 'onlineSavingsMsg'
Learn More

Benperidol, TRC

Product Code. 30011535
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011535 500 mg 500mg
30010918 250 mg 250mg
30007862 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011535 Supplier Toronto Research Chemicals Supplier No. TRCB170010500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2062-84-2
Molecular Formula C22 H24 F N3 O2
Molecular Weight (g/mol) 381.44
Synonym 1-[1-[4-(4-Fluorophenyl)-4-oxobutyl]-4-piperidinyl]-1,3-dihydro-2H-benzimidazol-2-one, 1-[1-[3-(p-Fluorobenzoyl)propyl]-4-piperidyl]-2-benzimidazolinone, 1-(1-[4-(p-Fluorophenyl)-4-oxobutyl]piperidin-4-yl)-2-benzimidazolinone, 1-[1-[3-(p-Fluorobenzoyl)propyl]-4-piperidyl]-2-benzimidazolinone, Anquil, Benperidol, Benzoperidol, Benzperidol, CB 8089, Frenactil, Frenactyl, Glianimon, McN-JR 4584, NSC 170982, R 4584, R 4730
IUPAC Name 3-[1-[4-(4-fluorophenyl)-4-oxobutyl]piperidin-4-yl]-1H-benzimidazol-2-one
SMILES Fc1ccc(cc1)C(=O)CCCN2CCC(CC2)N3C(=O)Nc4ccccc34

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C22H24FN3O2/c23-17-9-7-16(8-10-17)21(27)6-3-13-25-14-11-18(12-15-25)26-20-5-2-1-4-19(20)24-22(26)28/h1-2,4-5,7-10,18H,3,6,11-15H2,(H,24,28)
Formula Weight 381.1853
Chemical Name or Material Benperidol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.