Learn More
Buerkle™ PVC Benchtop Draining Rack with Collecting Pan
Product Code. 11921611 Shop All Bürkle Products
Change view
Click to view available options
Positions:
31
49
73
Size:
400 x 400 mm
500 x 500 mm
600 x 600 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Positions Size Unit Size
11921611 73 600 x 600 mm Each
11911611 31 400 x 400 mm Each
11931611 49 500 x 500 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11921611 Supplier Buerkle™ Supplier No. 96403060
Estimated Shipment: 21-10-2026
Buerkle™ PVC Benchtop Draining Rack with Collecting Pan is easy and flexible to set down on a table or working surface.
  • Two large supports with wide bases ensure sufficient stability
  • The collecting pan can simply be removed for emptying
  • The positions of the stable rods can be changed at any time
  • Flexible, can be used on both sides
  • Draining rack PVC, rods PP
  • Incl. spanner for the rods
TRUSTED_SUSTAINABILITY
Specifications
Color Blue/White
Material PVC, Polypropylene
Positions 73
Size 600 x 600 mm
Width (English) 23.6 in.
Height (English) 23.62 in.
Includes Spanner, collecting pan, 15 x 60 mm rods, 56 x 100 mm rods, 6 x 150 mm rods
Product Type Benchtop Draining Rack
Height (Metric) 600 mm
Width (Metric) 600 mm