Learn More
Fisherbrand™ Bench Stand for Water Cartridges
Product Code. 11954199 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Filtration and Purification Cartridges
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11954199 Water Filtration and Purification Cartridges Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11954199
  • ONLINE EXCLUSIVE PRICE
    627.30 EUR
    411.95 EUR / Pack of 1
    Save 215.35 EUR
    34% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
Accessory for mounting four water filtration and purification cartridges. Fisherbrand™ Bench Stands for Water Cartridges have an aluminum and plastic construction for sturdiness and durability.
  • With leveling feet and inlet faucet
  • Quick coupling for easy connection/disconnection
  • Comes with 15/21 tip and 20/37 adapter
Specifications
Cartridge Size 500 mm
For Use With (Equipment) Water Filtration and Purification Cartridges
Type Bench Stand
Purity Meter Included No