Learn More
Bel-Art™ Thermometer Holder
Product Code. 11347174
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
11347174 Polypropylene Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11347174 Supplier Bel-Art™ Supplier No. F182410000
  • ONLINE EXCLUSIVE PRICE
    21.35 EUR
    12.99 EUR / Pack of 3
    Save 8.36 EUR
    39% Off
Estimated Shipment: 22-12-2026
Easy-to-use, polypropylene clips hold thermometers and probes from 3.2 to 11 mm in diameter securely in place
  • Fits onto the rims of beakers or pitchers up to 1/4 in. thick
  • Autoclavable at 121°C (250°F)
  • Polypropylene
  • Autoclavable
Tekniske data
For Use With (Application) Secure and easy reading of thermometers
Material Polypropylene
Product Type Thermometer Holder
For Use With (Equipment) 3 to 10.9mm (0.12 to 0.43 in.) thermometers
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.