Learn More
Bel-Art™ SP Scienceware™ Sterileware™ Sterile Mini-Tongs
Product Code. 11362164
Change view
Click to view available options
Length (Metric):
11 cm
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11362164 11 cm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11362164 Supplier Bel-Art™ Supplier No. F379440000
  • ONLINE EXCLUSIVE PRICE
    69.40 EUR
    43.28 EUR / Pack of 25
    Save 26.12 EUR
    37% Off
View all promotions
Estimated Shipment: 20-01-2027
These small, lightweight tongs are ideal for sampling small quantities of material ranging from food to pharmaceuticals
These small, lightweight tongs are ideal for sampling small quantities of material ranging from food to pharmaceuticals
  • Ridged jaws securely hold wet or dry samples
  • One piece, sterile tongs are made from polystyrene plastic
  • Individually packaged, sterilized by gamma radiation, lot stamped for accurate lot tracing and shipped with a Certificate of Processing for Sterilization
Specifications
Product Type Mini Tongs
For Use With (Application) Ideal for sampling small quantities of material ranging from food to pharmaceuticals
Length (English) 4.25 in.
Tip Style Serrated
Length (Metric) 11 cm
Material Polystyrene