Learn More
Bel-Art™ Plate Reader
Description
- For examining cell monolayer, cell clones and for counting viral plaques or bacterial colonies
- Easy and safe reading of hemagglutination, hemagglutination inhibition, hemolysis and agglutination tests
- Clear deck for resting plates
- Large, adjustable mirror is perfect for viewing plate underside
- Mirror magnification is 3X actual plate size
- Made of polypropylene with a glass-clear acrylic deck for placement of plates and Petri dishes
Hemagglutination, Hemagglutination Inhibition, Hemolysis and Aglutination testing
Specifications
Specifications
| Product Type | Microtiter Plate Reader |
| Display Type | None |
| Magnification | 3X |
| Material | Polypropylene, Clear Glass Acrylic Deck |
| For Use With (Application) | Hemagglutination, hemagglutination inhibition, hemolysis and agglutination testing |
| For Use With (Equipment) | Microtiter plates |