Learn More
Bel-Art™ Fire Blanket
Shop All SP Scienceware Products
Click to view available options
Color:
Tan
Unit Size:
Each
Description
- For small Class A oil/grease fires and flare-ups
- May also be wrapped around the head and body for protection against flames and embers when fleeing a burning room, or for protection from splashing molten metal
- Woven fiberglass with a textured surface; all edges are serged with fireproof Kevlar™ thread
- Close weave greatly reduces the amount of atmospheric oxygen available to feed the fire
- Ppackaged in an instant access container that can be hung on the wall or stored in any accessible location
Specifications
Specifications
| Product Type | Fire Blanket |
| Color | Tan |
| Material | Woven Fiberglass |
| Dimensions (L x W) | 36 x 40 in. (91.4 x 101.6 cm) |
| For Use With (Application) | Ideal for class A light Oil and grease flare-ups |
| Length (English) | 36 in. |
| Length (Metric) | 91.4 cm |
| Voltage | 0/1 V |
| Width (English) | 40 in. |
| Width (Metric) | 101.6 cm |