Learn More
Bel-Art™ SP Scienceware™ Equilibrium Dialysis Cells
Product Code. 12733948
Change view
Click to view available options
Capacity (Metric):
1 mL
5 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12733948 5 mL Each
15204091 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12733948 Supplier Bel-Art™ Supplier No. H402610000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Feature stainless steel screws that tighten down on the dialysis membrane also acting as a gasket
  • Made from clear acrylic
  • Filler hole screws on each half cavity are fluoropolymer-coated stainless steel
  • Volume shown is for one half cell (double for total cell volume)
  • A red dot on one half of the equilibrium cell easily identifies the otherwise identical halves
Specifications
Product Type Dialysis Cell
Material Acrylic
Capacity (Metric) 5 mL
Color Transparent
Quantity Each
For Use With (Application) Stainless steel screws that tighten down on the dialysis membrane also acting as a gasket