Learn More
Bel-Art™ Disposal Benchtop Carton for Glass
Product Code. 11312204
Change view
Click to view available options
Autoclavable:
Not Autoclavable
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Autoclavable Unit Size
11312204 Not Autoclavable Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11312204 Supplier Bel-Art™ Supplier No. F246530002
  • ONLINE EXCLUSIVE PRICE
    85.75 EUR
    48.08 EUR / Pack of 6
    Save 37.67 EUR
    43% Off
Estimated Shipment: 22-12-2026
Coated with 2 mil thick, clear polyethylene bag to contain broken glass fragments and spilled liquids. Bel Art™ Disposal Benchtop Carton for Glass is easily closed for disposal.
Coated with 2 mil thick, clear polyethylene bag to contain broken glass fragments and spilled liquids. Bel Art™ SP Scienceware™ Disposal Benchtop Carton for Glass is easily closed for disposal.
  • Holds 5.5 kg (12 lb)
  • Packaged flat to save space and shipping costs and quickly assembled
  • Lined with a 2 mil, clear polyethylene bag to store broken glass fragments and leaked liquids
  • Printed in four languages: English, German, French, Spanish
  • Safe, puncture-resistant receptacles for broken glassware
  • Does not qualify as a DOT container
Specifications
Capacity (English) 2.6 gal.
Color Red
Dimensions (L x W x H) 20 x 20 x 25 cm
Autoclavable Not Autoclavable
Includes Closure, Graphics in four languages (English, German, French, Spanish)
For Use With (Application) Glass
Material Corrugated Cardboard
Product Type Safety Container