Learn More
Bel-Art™ Complete Fittings Assortment
Description
This generous assortment of 72 plastic fittings has everything needed for routine laboratory set-ups.
- Clear, hinge-top, polystyrene storage box makes it easy to find what you need
- Includes commonly used sizes of connector and stopper styles
- FDA grade, autoclavable polypropylene connectors: 15 Straight Tubing Connectors, 11 Stepped Tubing Connectors, 6 “T” Shaped Tubing Connectors, 10 Wye (Y) Tubing Connectors
- Polyethylene connectors: 3 pair Quick Disconnects
- Polyethylene stoppers: 24 Cup-Type
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Includes | Assortment of 72 plastic fittings |
| Color | Natural |
| Product Type | Fittings Assortment |