Learn More
Buerkle™ BeefSteaker Sampler
Product Code. 11816403
Change view
Click to view available options
Material:
Stainless Steel (V2A)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
11816403 Stainless Steel (V2A) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11816403 Supplier Buerkle™ Supplier No. 53881000
Estimated Shipment: 16-12-2026
Designed to take bacteriological samples from the surfaces of dead animals
  • For taking bacteriological samples from the surfaces of dead animals in slaughterhouses and in butcher shops (cattle, pigs, sheep, goats and horses)
  • Used to cut out a 25 mm round sample from animal skin
  • Stainless steel AISI 420 cutter allows the sample to be cut out easily
  • Can also be used for sampling in (histology, medicine, food and semi-luxury articles, cheese factories, agriculture)
  • Can be steam-sterilised or flamed
  • A hole in the crown prevents air compression and is used for ejecting the sample
TRUSTED_SUSTAINABILITY
Specifications
Product Type Meat Sampler
Material Stainless Steel (V2A)
Autoclavable Autoclavable
For Use With (Application) Foodstuffs, Pasty/highly viscous material, Surface sample, Histology, Medicine, Food and semi-luxury articles, Agriculture
Diameter (English) Core 0.98 in.
Diameter (English) Outer 1.06 in.
Diameter (Metric) Core 25 mm
Diameter (Metric) Outer 27 mm
Diameter (Metric) Outer Bottom 27 mm
Length (English) 7.32 in.
Length (Metric) 186 mm
Width (English) 3.15 in.
Width (Metric) 80 mm