Learn More
Beclomethasone 17-Valerate, TRC
Product Code. 30138727
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138727 100 mg 100mg
30004645 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138727 Supplier Toronto Research Chemicals Supplier No. TRCB131065100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 52619-18-8
Molecular Formula C27 H37 Cl O6
Molecular Weight (g/mol) 493.03
Synonym Pregna-1,4-diene-3,20-dione, 9-chloro-11,21-dihydroxy-16-methyl-17-[(1-oxopentyl)oxy]-, (11β,16β)- (9CI), (11β,16β)-9-Chloro-11,21-dihydroxy-16-methyl-17-[(1-oxopentyl)oxy]pregna-1,4-diene-3,20-dione, 9-Chloro-11β,21-dihydroxy-16β-methyl-3,20-dioxopregna1,4-dien-17-yl Pentanoate, Beclometasone 17-Valerate, Beclomethasone 17-Valerate, Betamethasone Valerate Imp. H (EP)
IUPAC Name [(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] pentanoate
SMILES CCCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C)C(=O)CO
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C27H37ClO6/c1-5-6-7-23(33)34-27(22(32)15-29)16(2)12-20-19-9-8-17-13-18(30)10-11-24(17,3)26(19,28)21(31)14-25(20,27)4/h10-11,13,16,19-21,29,31H,5-9,12,14-15H2,1-4H3/t16-,19-,20-,21-,24-,25-,26-,27-/m0/s1
Formula Weight 492.2279
Percent Purity >95%
Chemical Name or Material Beclomethasone 17-Valerate
Grade HPLC