Learn More
Pinnacle Brush™ Beaker Brushes
Product Code. 12811934
Change view
Click to view available options
Length (Metric):
26 cm
27 cm
36.5 cm
39 cm
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12811934 27 cm Pack of 10
12398318 26 cm Pack of 10
12821934 39 cm Pack of 10
12851934 36.5 cm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12811934 Supplier Pinnacle Brush™ Supplier No. T309
  • ONLINE EXCLUSIVE PRICE
    124.00 EUR
    106.80 EUR / Pack of 10
    Save 17.20 EUR
    13% Off
View all promotions
Estimated Shipment: 15-02-2027
Constructed on galvanised steel wire handle. Pinnacle Brush™ Beaker Brushes have nylon bristles for gentle cleaning.
  • Galvanized steel handle
  • Nylon bristles
Specifications
Product Type Bottle Brush
Handle Material Metal
Diameter (Metric) 4.5 cm
Length (Metric) 27 cm
Length (Metric) Brush 9.5 cm
Length (Metric) Handle 17.5 cm
Quantity 10 Pk.
Size Brush Circumference: 14 cm
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.